Learn More
BOLA Tube Fittings, Elbow
Product Code. p-200110024
Click to view available options
Diameter (Metric):
4 mm
8 mm
12 mm
14 mm
16 mm
Unit Size:
Each
Product Code. 10545201
Brand: BOLA D50402
Only null left
Add to basket
Add to basket
Tube Fittings, Elbow PTFE, nut made of glass-fibre reinforced PTFE
  • Tube fitting elbow-shaped made of PTFE, two connections with nuts made of glass-fibre reinforced PTFE.
  • Universal chemical resistance, the product is only exposed to PTFE.
Specifications
Certifications/Compliance FDA
Diameter (Metric) 4 mm
Autoclavable Autoclavable
Color White
Flexibility Rigid