Learn More
Eppendorf™ MixMate Tube Holder
Shop All Eppendorf Products
Click to view available options
Tube Capacity:
0.2 mL
0.5 mL
2 mL
No. of Positions:
24
96
Unit Size:
Each
Description
- Tube Holders for Eppendor MixMate™
- The 3-in-1 mixer for plates, tubes and vortex mixing
- High speed instrument guarantees complete mixing of your samples in seconds
- Outstanding 2D Mix-Control provides optimized mixing with anti-spill technology
Specifications
Specifications
| Tube Capacity | 0.2 mL |
| Product Type | Tube Holder |
| For Use With (Equipment) | 96 x 0.2mL PCR tubes, pcr strips/96-well pcr plates semi-skirted/unskirted, MixMate vortex mixer |
| No. of Positions | 96 |