Learn More
Greiner Bio-One Tube, Polypropylene, Round Bottom, Non-Sterile
Product Code. 10353821
Click to view available options
Capacity (Metric):
5 mL
Unit Size:
Pack of 2000
Product Code. 10353821
Brand: Greiner Bio-One 115201
Only null left
Add to basket
Add to basket
Suitable for a variety of purposes and featuring good thermal, chemical, and mechanical stability.
  • Polypropylene tubes have particularly good thermal, chemical and mechanical stability
  • Material is ideally suited for the storage of chemical and biological samples
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 12 mm
Capacity (Metric) 5 mL
Material Polypropylene
Length (Metric) 75 mm
Quantity 250/Pk.