Learn More
Masterflex™ Tubing Adapter NPT (F) Polypropylene
Ideal for general-purpose and single-use applications, to connect tubing with rest of system
Brand: Masterflex™ 31200-11
Additional Details : Weight : 0.01880kg
Description
- Determine which connector type is needed for your application by contacting free technical application assistance
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Always in stock—most materials and configurations available to ship same day
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
- All fittings are compatible with Tri-Clamp™ connections
- Excellent chemical resistance; low leachable characteristics
Specifications
Vacuum Tubing | |
Masterflex L/S and I/P sanitary tubing connection | |
1/4 in. | |
Polypropylene |
Tubing Adapter; Polypropylene (PP), 3/4 in. mini to 1/4 in. NPT (F) | |
Sanitary x NPT(F) | |
Connection Type: 3/4 in. mini to 1/4 in. NPT(F) |