missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Isoflex Laboratory Hose
Kartell™ Isoflex Laboratory Hose is a transparent, non-toxic hose that features markings ever meter for ease of use.
Brand: Kartell™ 0176000
Additional Details : Weight : 0.10800kg
Description
- Autoclavable PVC hoses
- Transparent
- Non toxic
- Non ageing
- Non oxiding
- Supplied in 20 meter rolls packed in PE bags
- The hoses is marked every meter for ease of use
- Good chemical resistance
- Good strong and weak acid resistance
- Good strong and weak alkali resistance
Specifications
Autoclavable | |
PVC | |
20 m | |
Transparent |
0.8 mm | |
2.4 mm | |
0.8 mm |