Learn More
Sartorius Stedim Biotech™ TuFlux Silicone Tubing
Highly resistant platinum-cured silicone tubing designed to facilitate fluid transfer in many pharmaceutical and biopharmaceutical applications such as media and buffer processing, filtration, fermentation and cell harvesting.
Brand: Sartorius Stedim Biotech™ FSA117425
Additional Details : Weight : 0.33000kg
Description
- Highly resistant platinum cured silicone tubing designed to facilitate fluid transfer in pharmaceutical and biopharmaceutical applications such as media and buffer processing, filtration, fermentation and cell harvesting.
- Can be mounted on Sartorius Stedim Biotech single-use assemblies or is available as non sterile tubing coils.
- Pure, clean, strong, and easy to handle.
- Low extractable profile.
Specifications
E.P. 3.1.9, USP <88> Class VI, ADCF | |
3.2 mm | |
6.4 mm | |
1.6 mm |
Autoclavable | |
Silicone (Platinum-Cured) | |
100 m | |
Autoclave, Gamma |