Learn More
Thermo Scientific™ Tulobuterol
beta-2-adrenergic receptor agonist
Brand:  Thermo Scientific™ J63975.03
 View more versions of this product
Product Code. 15456119
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 g
-30°C to -10°C
beta-2-adrenergic receptor agonist
C12H18ClNO
159°C to 163°C
White to Off-white
Crystalline Powder
41570-61-0
227.73g/mol