missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Tumor Necrosis Factor-alpha (78-96), human
379.00€ - 495.00€
Specifications
Content And Storage | Store at -20°C |
---|---|
Format | Solid |
Sensitivity | Light sensitive |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15427559
|
Thermo Scientific™
J66267.#0 |
5 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15437559
|
Thermo Scientific™
J66267.MA |
10 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Store at -20°C | |
Solid | |
Light sensitive |