Learn More
Buerkle™ Turn 'n' Spray™ Spray Bottles with Overhead Valve
10.16 EUR valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Use promo code "23267" to get your promotional price.
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Height (Metric):
220 mm
240 mm
295 mm
Neck ID (Metric):
17.5 mm
18 mm
Description
Turn'n'Spray features a special technology allowing you to spray in any direction and reach hard-to-access places easily and without straining.
- Uninterrupted spraying at any angle
- Parts in contact with contents made of PP, PE, silicon and stainless steel
- Nozzle diameter 0.6mm, nozzle is continuously adjustable
- Amount sprayed per stroke: 1.2mL ± 0.1mL
Specifications
Height (Metric) | 220 mm |
Capacity (Metric) | 250 mL |
Includes | Overhead valve |
Material | Polyethylene, Polypropylene, Silicon, Stainless Steel |
Neck ID (Metric) | 18 mm |