Learn More
Labplas Twirl'EM™ Large Format Sampling Bags with Tabs
Product Code. 16336378
Click to view available options
Capacity (Metric):
6.75 L
Unit Size:
Pack of 250
Product Code. 16336378
Brand: Labplas EDL41218
Only null left
Add to basket
Add to basket
Polyethylene sampling bags with large openings
  • Large sized
  • Contaminant-free
  • Virgin polyethylene
  • Large bag mouth opening
  • Easy to use pull tabs
Specifications
Description Regular Tabs
Capacity (Metric) 6.75 L
Closure Type 2 Flat Wires with Tabs
Material Polyethylene
Sterility Sterile
Thickness (English) 4 mil
Dimensions (L x W) 18 x 12 in. (457 x 305mm)
Length (English) 18 in.
Length (Metric) 457 mm
Width (English) 12 in.
Width (Metric) 305 mm