Learn More
Hamilton™ Two-Hub Assembled Tubing Assemblies
Chemically inert for fluid handling in the laboratory.
Brand:  Hamilton™ 88917
Additional Details : Weight : 0.10000kg
Product Code. 10281522
  • 51.00€ / Pack of 2
Estimated Shipment: 13-05-2024
to see stock.
Description
Description
  • 304.8mm in length (12 in.)
  • 18 gauge
Specifications
Specifications
FEP
18 G
304.8 mm
Tubing