Learn More
DuPont™ Tychem™ 6000 F with Socks
Shop All DuPont Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 20
Description
- Made from a lightweight and durable fabric (< 500 g per garment) consisting of a proprietary barrier film laminated to a heavy duty Tyvek™ substrate
- Offer excellent chemical permeation protection to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards, and certain chemical warfare agents
- Certified according to Regulation (EU) 2016/425
- Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B
- EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination)
- Antistatic treatment (EN 1149-5) on inside
- Stitched and over-taped seams with barrier tape for protection and strength
- Self-adhesive double zipper flap closure system for higher protection
- Used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications
Specifications
Specifications
| Color | Gray |
| Size | 2X-Large |
| Ankle Style | Elastic, Attached socks, Boot overflaps |
| Closure Type | Double zipper with self-adhesive flap and chin flap |
| Cuff Style | Thumb Loop |
| Description | Individually Packed |
| Disposable | Single-use |
| Gender | Unisex |
| Neck Style | Hooded |
| Quantity | 20/Cs. |
| Show More |