Learn More
Masterflex™ Tygon™ 3 Stop Hydrocarbon Tubing
Tailored for petro-chemicals and resists swelling and cracking. Tygon™ 3 Stop Hydrocarbon Tubing is also UV and Ozone resistant.
Brand: Masterflex™ 95605-46
Additional Details : Weight : 0.05000kg
Description
- Resists embrittlement
- Compatible with most hydrocarbons
- Resistant to swelling
- Highly flexible, easy to install
- Specifically developed for fuels and lubricants
- Ozone and UV light resistant
Specifications
Purple/Purple/Orange | |
Tygon F4040 | |
12 Pack |
2.54 mm | |
0.85 mm |