Learn More
Masterflex™ Tygon SE–200 Tubing
Brand:  Masterflex™ 95713-30
Additional Details : Weight : 2.00000kg
 View more versions of this product
Product Code. 15122045
  • 595.00€ / 15m
In Stock
to see stock.
Add to basket
Specifications
FDA Compliant for contact food products
Tygon SE-200
15.24 m
Chemical and pharmaceutical processing, beverage dispensing, paint and solvent production, semiconductor processing
1 Each
Transparent
74°C
9.6 mm
9.6 mm
2.3 mm
-45°C
Sterile
50 psi