Learn More
Justrite™ Type I Polyethylene Safety Shelf Can
Brand:  Justrite™ 14545Z
Additional Details : Weight : 3.50000kg
Product Code. 13443810
  • 349.00€ / Each
Estimated Shipment: 20-06-2024
to see stock.
Specifications
Shelf Can
18.93 L
Polyethylene
292 mm
FM, NFPA, OSHA Certified
Red
432 mm