Learn More
Dostmann Electronic™ Type K Thermo Couple Immersion
Brand:  Dostmann Electronic™ 6010-1006
Additional Details : Weight : 0.80000kg
Product Code. 10015961
  • 110.00€ / Each
Estimated Shipment: 26-07-2024
to see stock.
Add to basket
Récupel-bijdrage uitgesloten
Description
Description
Order Info
LengthMetric
Specifications
Specifications
Thermocouple Immersion
Class 1
300 mm
PVC
-200°C to +1100°C