missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TYRO3 (RSE) Recombinant Human Protein

TYRO3 is a receptor tyrosine protein kinase.

Brand:  Invitrogen™ PR7480A

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15408603

  • 1800.00€ / 100µg
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TYRO3 has been shown to be involved in signal transduction and cell adhesion.

Specifications

Specifications

NP_006284
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
100μg
RUO - research use only
TYRO3 (RSE), DKT, BRT, TIF
Human
GST Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
76.6kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
AXL Kinase Family
Recombinant
Baculovirus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions