missing translation for 'onlineSavingsMsg'
Learn More

uvex U-Fit Lite Nitrile Chemical Protection Gloves

Product Code. p-7236918 Shop All Uvex Products
Click to view available options
Size:
7
8
9
10
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15397142

Brand: uvex 6059709

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Uvex U-Fit Lite disposable safety gloves are extremly lightweight, while still providing good protection against dirt and oil. Gloves are touchscreen compatible and ideal for light precision assembly work.

  • Lightweight, durable disposable NBR gloves
  • Texturized fingertips for enhanced grip
  • Reliable spray protection when handling chemicals such as acids, alkalis, solids, or aqueous saline solutions
  • Silicone-free according to the imprint test
  • Touchscreen compatible
  • Food safe
TRUSTED_SUSTAINABILITY

Specifications

Accelerator Free Yes
Color Blue
Disposable Single-use
Exterior Finish Smooth
Latex Free Yes
Material Nitrile Rubber
Palm Thickness (Metric) 0.08 mm
Powdered Powder Free
Product Type Nitrile Gloves
Size 9
Sterility Non-sterile
EN ISO 374-1 Type C
EN ISO 374-1 CHEMICAL LETTER CODES K
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection
Quantity Box of 100
Series U-Fit Lite
Length (Metric) 24 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.