missing translation for 'onlineSavingsMsg'
Learn More

uvex U-Fit Strong N2000 Chemical Protection Gloves

Product Code. 17210521 Shop All Uvex Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17210521

Brand: uvex 6096209

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Reinforced, disposable nitrile rubber safety gloves provide protection against chemicals and splashes, with a high degree of mechanical resistance.

  • Reinforced, disposable nitrile rubber safety gloves
  • Textured fingertips for enhanced grip capabilities
  • Resistant to grease, mineral oils, and a wide variety of chemicals
  • Silicone-free according to imprint test
  • Ideal for cleaning, laboratory work, and the food industry
TRUSTED_SUSTAINABILITY

Specifications

Applications Laboratory
Color Blue
Disposable Single-use
Latex Free Yes
Material Nitrile Rubber
Product Type Chemical-resistant Gloves
Size Large
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKLOPST
For Use With (Application) Chemical Handling, Cleaning, Food Industry, Laboratory Work, and Painting
Gender Unisex
Series U-Fit Strong N2000
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.