Learn More
Poulten Selfe™ U-Tube Viscometer
Click to view available options
Viscosity Range:
6 to 30 cSt
Size:
C
Unit Size:
Each
Description
- PSL calibrated viscometers are supplied with UKAS certificates providing an official assurance of the competence of the calibration work carried out. UKAS certificates of calibration, which satisfy the requirements of ISO 9000, are issued by the PSL Calibration Laboratory, which is a National Measurement Accreditation Service accredited laboratory.
- Type BS/U, IP71.Suitable for transparent liquids. Nominal overall length 300mm. Approx. sample size 13mL to 40mL.
Specifications
Specifications
| Product Type | U-Tube Viscometer |
| Viscosity Range | 6 to 30 cSt |
| Certifications/Compliance | UKAS, ISO 9000 |
| Size | C |
| Volume (Metric) | 18 mL |