missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ UC 112

Product Code. 18750509
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18750509

Brand: Tocris Bioscience™ 5251/10

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

IAP and XIAP inhibitor; antitumor

UC 112 is an iAP inhibitor (IC50 values from 0.7 - 3.4 μM); inhibits cell growth in multiple cancer cell lines and in a melanoma xenograft model in vivo. Also suppresses X-linked inhibitor of apoptosis protein (XIAP) and survivin levels. Inhibits the growth of P-glycoproteins and activates caspase-3/7 and caspase-9.
TRUSTED_SUSTAINABILITY

Specifications

Purity >98%
Quantity 10 mg
Product Type UC 112
Molecular Weight (g/mol) 348.44
Inhibitors UC 112

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.