missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ UC 112
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
UC 112 is an iAP inhibitor (IC50 values from 0.7 - 3.4 μM); inhibits cell growth in multiple cancer cell lines and in a melanoma xenograft model in vivo. Also suppresses X-linked inhibitor of apoptosis protein (XIAP) and survivin levels. Inhibits the growth of P-glycoproteins and activates caspase-3/7 and caspase-9.
Specifications
Specifications
| Purity | >98% |
| Quantity | 10 mg |
| Product Type | UC 112 |
| Molecular Weight (g/mol) | 348.44 |
| Inhibitors | UC 112 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction