missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, Ultra-Pure, Chem-Lab
Click to view available options
Quantity:
2.5 L
5 L
10 L
Specifications
pH | 7 |
Color | Colorless |
Melting Point | 0°C |
Boiling Point | 100°C |
Formula Weight | 18.016g/mol |
Grade | Ultra Pure |
Vapor Pressure | 17.5mm Hg at 20°C |
Filtered Through | LF < 1μS – o0.4μm, UV filtrated |
Density | 1.00g/mL |
Quantity | 2.5 L |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'