missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ UltraPool™ HLM 150, Mixed Gender
Click to view available options
Quantity:
0.5 mL
Description
- Large pool of 150 donors, 300 alleles for diploid genes, with an equal donor mix on a per milligram microsomal protein basis
- Provides the best representation of an “average patient” drug metabolism activity, including known CYP polymorphism
- Statistically modeled so that lot-to-lot variability represented as coefficient of variation (CV) is <10% (mean) for the more variable CYP enzyme (CYP2C19), and <5% (mean) for the key drug metabolizing CYPs (CYP1A2, CYP2C9, CYP2D6, and CYP3A4).
- Fully characterized for many important cytochrome P450, UGTs and other enzymes.
- Large lot size available, with matching S9 and cytosol products
- Tissues are sourced from US-based Organ Procurement Organizations, meeting high ethical standards with strict traceability
- Manufactured by adhering to high safety standards including serological testing prior to acceptance, followed by PCR testing for HIV I/II, Hepatitis B, Hepatitis C, HTLV 1/2 (human T-cell leukemia virus) pathogens, and CMV (cytomegalovirus)
- Protein Concentration: 20 mg/mL in 250 mM Sucrose
- Shelf life: N/A (this product is stable for more than 10 years when stored properly)
Specifications
Specifications
| Content And Storage | -80°C |
| Quantity | 0.5 mL |
| Species | Human |
| Source | US-based Organ Procurement Organizations, meeting high ethical standards with strict traceability |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction