Learn More
Qsonica Ultrasonic Homogenizer Converters
Shop All Fisherbrand Products
Click to view available options
Product Type:
Converter Cable
Unit Size:
Each
Description
- For use with Microson™ Model XL2000 or Sonicator™ 3000 Ultrasonic Homgenizers
- Transforms electrical signal into mechanical vibrations
Specifications
Specifications
| Product Type | Converter Cable |
| Length (English) | 72 in. |
| Length (Metric) | 182.88 cm |
| For Use With (Application) | Ideal for transform electrical signal into mechanical vibration |
| For Use With (Equipment) | Q500 and Q700 Sonicator™ |