Learn More
DiscoverX UNC 0638
Product Code. 17322479
Encompass
Click to view available options
:
10 mg
Product Code. 17322479
Brand: DiscoverX 921150
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
UNC 0638 is a cell permeable synthetic small molecule inhibitor of the G9a and GLP histone lysine methyltransferase enzymes (IC50 of <15nM and 19nM, respectively), which contribute to early embryogenesis, genomic imprinting, and lymphocyte development. >1000-fold selective vs other HMTases.
UNC 0638 is a cell permeable synthetic small molecule inhibitor of the G9a and GLP histone lysine methyltransferase enzymes, which contribute to early embryogenesis, genomic imprinting, and lymphocyte development. >1000-fold selective vs other HMTases.
Specifications
Quantity 10 mg