Learn More
DiscoverX UNC 0638
Click to view available options
:
10 mg
Description
UNC 0638 is a cell permeable synthetic small molecule inhibitor of the G9a and GLP histone lysine methyltransferase enzymes, which contribute to early embryogenesis, genomic imprinting, and lymphocyte development. >1000-fold selective vs other HMTases.
Specifications
Specifications
| Quantity | 10 mg |