missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ UNC 6934
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
UNC 6934 binds to the N-terminal proline-tryptophan-tryptophan-proline (PWWP) domain of nuclear receptor-binding SET domain-containing 2 (NSD2). UNC 6934 inhibits PWWP1 interaction with nucleosomal H3K36me2 (IC50 = 104 nM) by occupying the canonical H3K36me2-binding pocket of PWWP1. It selectively binds endogenous NSD2 in cells and causes accumulation of endogenous NSD2 in the nucleolus.
Specifications
Specifications
| Chemical Name or Material | N-Cyclopropyl-3,4-dihydro-3-oxo-N-[[4-[(4-pyrimidinylamino)carbonyl]phenyl]methyl]-2H-1,4-benzoxazine-7-carboxamide |
| CAS | 2561494-77-5 |
| Quantity | 10 mg |
| Target | Lysine Methyltransferase Inhibitors |
| Molecular Formula | C24H21N5O4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction