Learn More
KARTELL Unequal Straight Connector
The Unequal Straight Connector has different diameters on the inlet and outlet to connect hoses of different sizes.
Brand:  KARTELL 0087700
Additional Details : Weight : 0.01000kg
Product Code. 12503317
  • 153.00€ / Pack of 100
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
Description
Description
  • Inlet and outlet have different diameters.
  • Autoclavable.
  • Dimensions are shown as both nominal and exact as elasticity of hoses will influence the choice of size.
  • Use hoses selector Art.1759 to determine ''grip''. This will then give equivalent crest dimension for the particular connector concerned.
  • TESTED ACCORDING TO REACH REGULATION 1907/2006 ANNEX XVII.
  • MATERIAL: PP
Specifications
Specifications
Tapered Connector
1.6, 4.6 mm
0.16/0.24/0.31 in.
White
Autoclavable
Polypropylene
4/6/8 mm