Learn More
KARTELL Unequal Straight Connector
The Unequal Straight Connector has different diameters on the inlet and outlet to connect hoses of different sizes.
Brand: KARTELL 0087700
Additional Details : Weight : 0.01000kg
Description
- Inlet and outlet have different diameters.
- Autoclavable.
- Dimensions are shown as both nominal and exact as elasticity of hoses will influence the choice of size.
- Use hoses selector Art.1759 to determine ''grip''. This will then give equivalent crest dimension for the particular connector concerned.
- TESTED ACCORDING TO REACH REGULATION 1907/2006 ANNEX XVII.
- MATERIAL: PP
Specifications
Tapered Connector | |
1.6, 4.6 mm | |
0.16/0.24/0.31 in. | |
White |
Autoclavable | |
Polypropylene | |
4/6/8 mm |