Learn More
Abeba™ UNI6 1762 Shoes
Product Code. 15657657
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15657657
Brand: Abeba™ 1762/35
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Steel toe-cap provides extra protection. Special lining with silver ions absorbs resperation allowing the foot to breathe.
  • CE EN ISO 20345:2011 S1 SRC
  • Functional leather with honeycomb pattern
  • Blue
  • Steel toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350122)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Scratch-proof
  • **Size 35 with replaceable acc Wave insole (art. 3576)
  • Water-repellent
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S1 SRC
Color Blue
Material Leather
Size 35
Sole Features Slip-resistant
Sole Material Polyurethane
Style 1762
Toe Material Steel