Learn More
Abeba™ UNI6 31782 Shoes
Product Code. 15639257
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15639257
Brand: Abeba™ 31782/46
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Steel toe-cap provides extra protection. Special lining with silver ions absorbs resperation allowing the foot to breathe.
  • CE EN ISO 20345:2011 S2 SRC
  • Microfiber
  • Gray/Blue
  • Steel toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350122)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • **Size 35 with replaceable acc Wave insole (art. 3576)
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S2 SRC
Color Gray, Blue
Material Microfiber
Size 46
Sole Features Slip-resistant
Sole Material Polyurethane
Style 31782
Toe Material Steel