Learn More
Abeba™ UNI6 31785 Shoes
Steel toe-cap provides extra protection. Special lining with silver ions absorbs resperation allowing the foot to breathe.
Brand:  Abeba™ 31785/39
Additional Details : Weight : 1.12000kg
 View more versions of this product
Product Code. 15689287
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20345:2011 S2 SRC
  • Microfiber
  • Gray/Black
  • Steel toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350122)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • **Size 35 with replaceable acc Wave insole (art. 3576)
Specifications
Specifications
CE EN ISO 20345:2011 S2 SRC
Microfiber
Slip-resistant
31785
Gray, Black
39
Polyurethane
Steel