Learn More
Abeba™ UNI6 36701 Shoes
ESD dissipative shoe includes special lining with silver ions that absorbs resperation which allows the foot to breathe.
Brand:  Abeba™ 36701/48
Additional Details : Weight : 0.95000kg
 View more versions of this product
Product Code. 15640428
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20347:2012 O1 FO SRC
  • Velours with breathable textile inlays
  • Black
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • Instep area with lacing
  • Reflective material
Specifications
Specifications
CE EN ISO 20347:2012 O1 FO SRC
Microfiber
48
Polyurethane
Black
Velour
Slip-resistant
36701