Learn More
Abeba™ UNI6 36721 Shoes
ESD dissipative shoe includes special lining with silver ions that absorbs resperation which allows the foot to breathe.
124.22€
Specifications
Certifications/Compliance CE EN ISO 20347:2012 O1 FO SRC
Color Black
Material Velour
Sole Features Slip-resistant
Sole Material Polyurethane
View More Specs
Products 14
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15630458
View Documents Promotion Details
Abeba™
36721/35
35
124.22€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15640458
View Documents Promotion Details
Abeba™
36721/36
36
124.22€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15650458
View Documents Promotion Details
Abeba™
36721/37
37
124.22€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15660458
View Documents Promotion Details
Abeba™
36721/38
38
124.22€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15670458
View Documents Promotion Details
Abeba™
36721/39
39
124.22€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15680458
View Documents Promotion Details
Abeba™
36721/40
40
124.22€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15690458
View Documents Promotion Details
Abeba™
36721/41
41
124.22€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15600468
View Documents Promotion Details
Abeba™
36721/42
42
124.22€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15610468
View Documents Promotion Details
Abeba™
36721/43
43
124.22€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15620468
View Documents Promotion Details
Abeba™
36721/44
44
124.22€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15630468
View Documents Promotion Details
Abeba™
36721/45
45
124.22€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15640468
View Documents Promotion Details
Abeba™
36721/46
46
124.22€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15650468
View Documents Promotion Details
Abeba™
36721/47
47
124.22€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
15660468
View Documents Promotion Details
Abeba™
36721/48
48
124.22€
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 O1 FO SRC
  • Velours with breathable textile inlays
  • Black
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
Specifications
Specifications
CE EN ISO 20347:2012 O1 FO SRC
Velour
Polyurethane
Black
Slip-resistant
36721