Learn More
Abeba™ UNI6 36750 Shoes
ESD dissipative shoe includes special lining with silver ions that absorbs resperation which allows the foot to breathe.
Brand:  Abeba™ 36750/41
Additional Details : Weight : 0.95000kg
 View more versions of this product
Product Code. 15610508
  • 125.77€ / 1 pair
Estimated Shipment: 30-05-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 O2 FO SRC
  • Smooth leather white
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • Instep area with lacing
Specifications
Specifications
CE EN ISO 20347:2012 O2 FO SRC
Microfiber
41
Polyurethane
White
Leather
Slip-resistant
36750