Learn More
Abeba™ UNI6 36750 Shoes
ESD dissipative shoe includes special lining with silver ions that absorbs resperation which allows the foot to breathe.
Brand: Abeba™ 36750/36
Additional Details : Weight : 0.95000kg
Description
- CE EN ISO 20347:2012 O2 FO SRC
- Smooth leather white
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable Soft Comfort insole M (article no. 350112)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Lining microfiber, respiration absorbing
- Instep area with lacing
Specifications
CE EN ISO 20347:2012 O2 FO SRC | |
Microfiber | |
36 | |
Polyurethane |
White | |
Leather | |
Slip-resistant | |
36750 |