Learn More
Abeba™ UNI6 36750 Shoes
Product Code. 15660508
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15660508
Brand: Abeba™ 36750/46
Only null left
Add to basket
Add to basket
ESD dissipative shoe includes special lining with silver ions that absorbs resperation which allows the foot to breathe.
  • CE EN ISO 20347:2012 O2 FO SRC
  • Smooth leather white
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • Instep area with lacing
Specifications
Certifications/Compliance CE EN ISO 20347:2012 O2 FO SRC
Color White
Liner Material Microfiber
Material Leather
Size 46
Sole Features Slip-resistant
Sole Material Polyurethane
Style 36750