Learn More
Abeba™ UNI6 36750 Shoes
missing translation for 'orderingAttributeHoverText'
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
missing translation for 'unitSize'
1 pair
Description
- CE EN ISO 20347:2012 O2 FO SRC
- Smooth leather white
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable Soft Comfort insole M (article no. 350112)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Lining microfiber, respiration absorbing
- Instep area with lacing
Specifications
Specifications
| Certifications/Compliance | CE EN ISO 20347:2012 O2 FO SRC |
| Color | White |
| Liner Material | Microfiber |
| Material | Leather |
| Size | 48 |
| Sole Features | Slip-resistant |
| Sole Material | Polyurethane |
| Style | 36750 |