Learn More
Abeba™ UNI6 36763 Shoes
ESD dissipative shoe includes special lining with silver ions that absorbs resperation which allows the foot to breathe.
Brand: Abeba™ 36763/46
Additional Details : Weight : 0.98000kg
Description
- CE EN ISO 20347:2012 O1 FO SRC
- Functional leather
- Dark blue
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable Soft Comfort insole M (article no. 350112)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Scratch-proof
- Water-repellent
Spécification
CE EN ISO 20347:2012 O1 FO SRC | |
Leather | |
Slip-resistant | |
36763 |
Blue | |
46 | |
Polyurethane |