Learn More
Abeba™ UNI6 36763 Shoes
ESD dissipative shoe includes special lining with silver ions that absorbs resperation which allows the foot to breathe.
Brand:  Abeba™ 36763/35
Additional Details : Weight : 0.98000kg
 View more versions of this product
Product Code. 15690578
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20347:2012 O1 FO SRC
  • Functional leather
  • Dark blue
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Scratch-proof
  • Water-repellent
Specifications
Specifications
CE EN ISO 20347:2012 O1 FO SRC
Leather
Slip-resistant
36763
Blue
35
Polyurethane