Learn More
Abeba™ UNI6 6731 Shoes
Special lining with silver ions absorbs resperation allowing the foot to breathe. Sanitized™ treated.
Brand: Abeba™ 6731/40
Additional Details : Weight : 0.98000kg
Description
- CE EN ISO 20347:2012 O1 FO SRC
- Smooth leather, perforated
- Black
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable Soft Comfort insole M (article no. 350112)
- Sanitized treated
- Antistatic
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
Specifications
CE EN ISO 20347:2012 O1 FO SRC | |
Leather | |
Slip-resistant | |
6731 |
Black | |
40 | |
Polyurethane |