Learn More
Abeba™ UNI6 6731 Shoes
Special lining with silver ions absorbs resperation allowing the foot to breathe. Sanitized™ treated.
Brand:  Abeba™ 6731/46
Additional Details : Weight : 0.98000kg
 View more versions of this product
Product Code. 15662068
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20347:2012 O1 FO SRC
  • Smooth leather, perforated
  • Black
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
Specifications
Specifications
CE EN ISO 20347:2012 O1 FO SRC
Leather
Slip-resistant
6731
Black
46
Polyurethane