Learn More
Abeba™ UNI6 6785 Shoes
Special lining with silver ions absorbs resperation allowing the foot to breathe. Sanitized™ treated.
Brand: Abeba™ 6785/41
Additional Details : Weight : 0.95000kg
Description
- CE EN ISO 20347:2012 O2 FO SRC
- Microfiber
- Gray/Black
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable Soft Comfort insole M (article no. 350112)
- Sanitized treated
- Antistatic
- Breathable lining with Silver ions
- Instep area with lacing
Specifications
CE EN ISO 20347:2012 O2 FO SRC | |
Microfiber | |
Slip-resistant | |
6785 |
Gray, Black | |
41 | |
Polyurethane |