Learn More
ABEBA™ Uni6 Black Safety Slip-On Shoe S2
Product Code. 16368021
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 16368021
Brand: ABEBA™ 31741/47
Only null left
Add to basket
Add to basket
ABEBA™ Uni6 Black Safety Slip-On Shoe S2 made of water and chemical resistant microfiber with a slip resistant PU/TPU outsole.
  • Comfort, safety and hygiene
  • Comply to EN ISO 20345 : 2012, S2, SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant PU/TPU outsole
  • Water and chemical resistant microfiber
  • Steel toe cap (200 joules)
  • Instep area with elastic band
  • Breathable and antibacterial lining with silver ions
  • Sanitized treatment
  • No side stitching
  • Removable insole Active comfort
Specifications
Certifications/Compliance CE EN ISO 20345:2012 S2 SRC ESD
Color Black
Liner Material Breathable silver ions lining
Lug Type Padded
Material Microfiber
Size 47
Sole Features Slip and abrasion resistant, ESD dissipative
Sole Material Polyurethane, Thermoplastic Polyurethane
Style Slip-on shoes
Toe Material Steel
Packaging Quantity 1 Pair
Certifications/Compliance (Europe) EN ISO 20345 : 2012; DIN EN 61340
Resistant To Water
Waterproof Yes