Learn More
Marienfeld Superior™ UniMark™ Uncoated Microscope Slides with Printed Marking Area
Product Code. 10285232
Click to view available options
Color:
Green
White
Edges:
90°, Ground
Cut
Unit Size:
Pack of 2500
Pack of 50
Product Code. 10285232
Brand: Marienfeld Superior™ 0704002
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Marienfeld Superior™ UniMark™ miscroscope slides have a printed area which can take notes printed by most types of printers and can be written with permanent markers. Different colors of the labelling area offer the possibility to distinct the preparations (by users, etc.)
  • Made of soda lime glass of third hydrolytic class
  • In compliance with DIN ISO 8037/1
  • Pre-cleaned, ready for use, autoclavable
  • With printed marking area in different colors
  • 50 pieces in a box, 50 boxes in a carton
  • For in-vitro diagnostic (IVD) applications according to IVD directive 98/79/EC, with CE-mark, recommended best-before-date and batch number for comprehensive information and traceability
Specifications
Length (Metric) 76 mm
Width (Metric) 26 mm
No. per Case 2500
No. per Pack 50
Edges 90°, Ground
Frosted Yes
Material Soda Lime Glass Type III, Hydrolytic Class
Precleaned Yes
Thickness (Metric) 1 mm
Type Microscope Slide
Color White
Certifications/Compliance CE Mark, DIN ISO 8037/1
Description UniMark™ Microscope Slides with printed marking area
Quantity 50 Pieces in Cardboard Box
Dimensions (L x W) 76 x 26 mm