Learn More
Erlab Uninterrupted Power Supply
To ensure your safety, the ERLAB Solis Premium is equipped with a UPS (uninterrupted power supply).
Brand: Erlab NORT000781
This item is not returnable.
View return policy
Description
- This device keeps the safety cabinet working properly for 10 minutes in the event of a power failure, allowing you to secure your handlings before the cabinet comes to a complete stop.