Learn More
Erlab Uninterrupted Power Supply
To ensure your safety, the ERLAB Solis Premium is equipped with a UPS (uninterrupted power supply).
Brand:  Erlab NORT000781
Product Code. 17776614
  • 552.00€ / Each
Description
  • This device keeps the safety cabinet working properly for 10 minutes in the event of a power failure, allowing you to secure your handlings before the cabinet comes to a complete stop.