Learn More
Smeg™ SB28 Universal Basket
Use this support with Smeg™ glassware washers. Smeg SB28 Universal Baskets have a capacity of 28 items and are made of stainless steel.
Brand:  Smeg™ SB28
Additional Details : Weight : 0.50000kg
Product Code. 11905574
  • 252.00€ / Each
Estimated Shipment: 17-06-2024
to see stock.
Add to basket
Description
Description
  • 28-position spring support to hold flasks, beakers etc.
  • For use with the standard baskets CS1-1 and CS2 and the trolley LM20
  • Recommended for flasks with a capacity of up to 1000mL
Specifications
Specifications
Used for supporting beakers during wash cycle
For Smeg 60cm trolley washer
1 Each