Learn More
Zeiss™ Universal Mounting Frame
Description
Compatible Microscopes:
- Axio Observer 3
- Axio Observer 5
- Axio Observer 7
- Axio Observer.A1
- Axio Observer.D1
- Axio Observer.D1 TIRF 3
- Axio Observer.Z1
- Axio Observer.Z1 TIRF 3
- Axio Vert.A1
- Axio Vert.A1 FL
- Axio Vert.A1 FL-LED
- Axio Vert.A1 MAT
- Axiovert 200
- Axiovert 200 M
- Axiovert 25
- Axiovert 25 C
- Axiovert 25 CFL
- Axiovert 40 C
- Axiovert 40 CFL
- Primovert
Specifications
Specifications
| Type | Universal Mounting Frame M-X |
| For Use With (Equipment) | Multitest dishes with foot print 124 to 133mm x 83 to 88mm and Petri dishes dia.: 87 to 92mm |
| Quantity | 1 Ea. |