Learn More
Macherey-Nagel™ Universal RP Guard Columns
Product Code. 15284668 Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric) Inner:
3 mm
Unit Size:
Each
Product Code. 15284668
Brand: Macherey-Nagel™ 728777.30
Only null left
Specifications
Length (Metric) 4 mm
Type RP Guard Column
Diameter (Metric) Inner 3 mm
Column Format Guard Column
For Use With (Application) HPLC