Learn More
Greiner Bio-One Universal Sterile Filter Tips for Gilson™ P1000
Prevents contamination with liquids and aerosols during pipetting process.
Brand:  Greiner Bio-One 740288
Additional Details : Weight : 1.62000kg
Product Code. 10449381
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • Minimizes danger of unwanted cross-contamination
  • Acts as reliable barrier against the transfer or aerosols into shaft of pipette
  • Can be used with all standard pipettes
  • Maintain tolerance limits stipulated for pipette tips i
Specifications
Specifications
1000 μL
Sterile
Irradiation
Yes
Filtered
White
Anthos™ New Line, Costar™ Onepette™, Eppendorf™ Research™, Thermo Scientific™ Finnpipette™ Digital, Gilson™ Pipetman™, Gilson™ Pipetman™ P200, Human™ Pipettes
Polypropylene