Learn More
Greiner Bio-One Universal Sterile Filter Tips With Graduation
Prevent contaminations with liquids and aerosols during pipetting process.
Brand:  Greiner Bio-One 739288
Additional Details : Weight : 1.33000kg
Product Code. 10524332
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Non-cytotoxic
  • Prevents aspiration of particles into interior of pipette
  • Minimizes cross-contamination
  • Ultra-micro filter made of hydrophobic polyethyl
Specifications
Specifications
200 μL
Sterile
Not Autoclavable
Irradiation
Yes
Filtered
Natural
−20° to 80°C
Anthos™ New Line, Costar™ Onepette™, Eppendorf™ Research™, Thermo Scientific™ Finnpipette™ Digital, Gilson™ Pipetman™, Gilson™ Pipetman™ P200, Human™ Pipettes
Polypropylene